missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trimethoxytoluene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1520118

 View more versions of this product

Catalog No. AAA1520118

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,4,5-Trimethoxytoluene
34°C to 38°C
115°C to 116°C (4 mmHg)
C10H14O3
MFCD00008397
1953791
KCIZTNZGSBSSRM-UHFFFAOYSA-N
1,2,3-trimethoxy-5-methylbenzene
80922
182.22
6443-69-2
1.082
74°C (165°F)
1.523
50 g
3,4,5-trimethoxytoluene, benzene, 1,2,3-trimethoxy-5-methyl, 5-methylpyrogallol trimethyl ether, 1,2,3-trimethoxy-5-methyl-benzene, toluene, 3,4,5-trimethoxy, 1,2,3-trimethoxy-5-methylbenzol, pubchem13665, acmc-209nm4, 3,4,5-trimethoxy-toluene, 3,4,5-trimethoxy toluene
CC1=CC(=C(C(=C1)OC)OC)OC
182.219
CHEBI:81354
98%

Chemical Identifiers

6443-69-2
182.219
KCIZTNZGSBSSRM-UHFFFAOYSA-N
80922
1,2,3-trimethoxy-5-methylbenzene
C10H14O3
MFCD00008397
3,4,5-trimethoxytoluene, benzene, 1,2,3-trimethoxy-5-methyl, 5-methylpyrogallol trimethyl ether, 1,2,3-trimethoxy-5-methyl-benzene, toluene, 3,4,5-trimethoxy, 1,2,3-trimethoxy-5-methylbenzol, pubchem13665, acmc-209nm4, 3,4,5-trimethoxy-toluene, 3,4,5-trimethoxy toluene
CHEBI:81354
CC1=CC(=C(C(=C1)OC)OC)OC
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 229-239-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only