missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Aminophenyl)propionic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 443600050

 View more versions of this product

Catalog No. AC443600050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-(4-Aminophenyl)propionic acid
Authentic
C9H11NO2
3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid
C1=CC(=CC=C1CCC(=O)O)N
165.19
165.19
2393-17-1
96% min. (HPLC)
5g
WXOHKMNWMKZMND-UHFFFAOYSA-N
3-(4-aminophenyl)propanoic acid
75451
97%

Chemical Identifiers

2393-17-1
165.19
3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid
3-(4-aminophenyl)propanoic acid
C9H11NO2
WXOHKMNWMKZMND-UHFFFAOYSA-N
75451
C1=CC(=CC=C1CCC(=O)O)N
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 219-245-1

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only