missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Di-O-acetyl-D-glucal, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6058503

 View more versions of this product

Catalog No. AAH6058503

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3,4-Di-O-acetyl-D-glucal
C10H14O6
1 g
LBVCUTFITQDWSU-BBBLOLIVSA-N
[(2R,3S,4R)-3-acetyloxy-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-4-yl] acetate
11064237
97%
82295-98-5
MFCD22989007
3,4-di-o-acetyl-d-glucal, 3-o,4-o-diacetyl-1,5-anhydro-2-deoxy-d-arabino-1-hexenitol, 2r,3s,4r-4-acetyloxy-2-hydroxymethyl-3,4-dihydro-2h-pyran-3-yl acetate
CC(=O)OC1C=COC(C1OC(=O)C)CO
230.216
230.21

Chemical Identifiers

82295-98-5
230.216
LBVCUTFITQDWSU-BBBLOLIVSA-N
11064237
CC(=O)OC1C=COC(C1OC(=O)C)CO
C10H14O6
MFCD22989007
3,4-di-o-acetyl-d-glucal, 3-o,4-o-diacetyl-1,5-anhydro-2-deoxy-d-arabino-1-hexenitol, 2r,3s,4r-4-acetyloxy-2-hydroxymethyl-3,4-dihydro-2h-pyran-3-yl acetate
[(2R,3S,4R)-3-acetyloxy-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-4-yl] acetate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions