missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Diacetyl-2,5-hexanedione 98.0+%, TCI America™
$356.68 - $1,066.73
Chemical Identifiers
CAS | 5027-32-7 |
---|---|
Molecular Formula | C10H14O4 |
Molecular Weight (g/mol) | 198.22 |
MDL Number | MFCD00031531 |
InChI Key | CSKRBHOAJUMOKJ-UHFFFAOYSA-N |
Synonym | 1,1,2,2-Tetraacetylethane, Bis(acetylacetone) |
PubChem CID | 78730 |
IUPAC Name | 3,4-diacetylhexane-2,5-dione |
SMILES | CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D4826200MG
|
TCI America
D4826200MG |
200 mg |
N/A
|
|
|||||
D48261G
|
TCI America
D48261G |
1 g |
N/A
|
|
|||||
Chemical Identifiers
5027-32-7 | |
198.22 | |
CSKRBHOAJUMOKJ-UHFFFAOYSA-N | |
78730 | |
CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O |
C10H14O4 | |
MFCD00031531 | |
1,1,2,2-Tetraacetylethane, Bis(acetylacetone) | |
3,4-diacetylhexane-2,5-dione |
Specifications
5027-32-7 | |
White-Yellow | |
C10H14O4 | |
200 mg | |
CSKRBHOAJUMOKJ-UHFFFAOYSA-N | |
3,4-diacetylhexane-2,5-dione | |
78730 | |
≥98.0% (GC) | |
3,4-Diacetyl-2,5-hexanedione |
193°C | |
122°C | |
MFCD00031531 | |
1,1,2,2-Tetraacetylethane, Bis(acetylacetone) | |
CC(=O)C(C(C(C)=O)C(C)=O)C(C)=O | |
198.22 | |
198.22 | |
Crystalline Powder |
Safety and Handling
TSCA : No