missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Diaminobenzamidoxime, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 1469816-70-3 |
---|---|
Molecular Formula | C7H10N4O |
Molecular Weight (g/mol) | 166.184 |
InChI Key | OPOKBHCWRPVYFH-UHFFFAOYSA-N |
Synonym | 3,4-diaminobenzamidoxime, 3,4-diamino-n-hydroxybenzimidamide |
PubChem CID | 86277480 |
IUPAC Name | 3,4-diamino-N'-hydroxybenzenecarboximidamide |
SMILES | C1=CC(=C(C=C1C(=NO)N)N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66495MD
|
Thermo Scientific Chemicals
H66495MD |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AAH6649503
|
Thermo Scientific Chemicals
H6649503 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1469816-70-3 | |
166.184 | |
3,4-diaminobenzamidoxime, 3,4-diamino-n-hydroxybenzimidamide | |
3,4-diamino-N'-hydroxybenzenecarboximidamide |
C7H10N4O | |
OPOKBHCWRPVYFH-UHFFFAOYSA-N | |
86277480 | |
C1=CC(=C(C=C1C(=NO)N)N)N |
Specifications
1469816-70-3 | |
250mg | |
OPOKBHCWRPVYFH-UHFFFAOYSA-N | |
3,4-diamino-N'-hydroxybenzenecarboximidamide | |
86277480 | |
97% |
C7H10N4O | |
3,4-diaminobenzamidoxime, 3,4-diamino-n-hydroxybenzimidamide | |
C1=CC(=C(C=C1C(=NO)N)N)N | |
166.184 | |
166.18 | |
3,4-Diaminobenzamidoxime |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO â Research Use Only