Learn More
3,4-Diaminobenzonitrile, 97%
CAS: 17626-40-3 | C7H7N3 | 133.15 g/mol
$71.49 - $893.25
Chemical Identifiers
| CAS | 17626-40-3 |
|---|---|
| Molecular Formula | C7H7N3 |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD00723901 |
| InChI Key | VWLLPPSBBHDXHK-UHFFFAOYSA-N |
| Synonym | 3,4-diamino-benzonitrile, benzonitrile, 3,4-diamino, 4-cyanobenzene-1,2-diamine, 3,4-diaminobenzenecarbonitrile, zlchem 619, pubchem2547, 3, 4-diaminobenzonitrile, 3,4 diamino benzonitrile, 3,4-diamino benzonitrile, acmc-209eb5 |
| PubChem CID | 737285 |
| IUPAC Name | 3,4-diaminobenzonitrile |
| SMILES | NC1=C(N)C=C(C=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6695403
|
Thermo Scientific Chemicals
H6695403 |
1 g |
Each for $71.49
|
|
|||||
|
AAH6695406
|
Thermo Scientific Chemicals
H6695406 |
5 g |
Each for $231.26
|
|
|||||
|
AAH6695414
|
Thermo Scientific Chemicals
H6695414 |
25 g |
Each for $893.25
|
|
|||||
Description
3,4-Diaminobenzonitrile is usually used like synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Diaminobenzonitrile is usually used like synthetic intermediate.
Solubility
Soluble in dichloromethane, methanol and ethyl acetate.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 17626-40-3 | |
| 133.15 | |
| VWLLPPSBBHDXHK-UHFFFAOYSA-N | |
| 737285 | |
| NC1=C(N)C=C(C=C1)C#N |
| C7H7N3 | |
| MFCD00723901 | |
| 3,4-diamino-benzonitrile, benzonitrile, 3,4-diamino, 4-cyanobenzene-1,2-diamine, 3,4-diaminobenzenecarbonitrile, zlchem 619, pubchem2547, 3, 4-diaminobenzonitrile, 3,4 diamino benzonitrile, 3,4-diamino benzonitrile, acmc-209eb5 | |
| 3,4-diaminobenzonitrile |
Specifications
| 17626-40-3 | |
| Brown | |
| MFCD00723901 | |
| UN3439 | |
| Soluble in dichloromethane,methanol and ethyl acetate. | |
| NC1=C(N)C=C(C=C1)C#N | |
| 133.15 | |
| 133.15 | |
| Solid |
| 140°C to 148°C | |
| C7H7N3 | |
| 1 g | |
| 3,4-diamino-benzonitrile, benzonitrile, 3,4-diamino, 4-cyanobenzene-1,2-diamine, 3,4-diaminobenzenecarbonitrile, zlchem 619, pubchem2547, 3, 4-diaminobenzonitrile, 3,4 diamino benzonitrile, 3,4-diamino benzonitrile, acmc-209eb5 | |
| VWLLPPSBBHDXHK-UHFFFAOYSA-N | |
| 3,4-diaminobenzonitrile | |
| 737285 | |
| 97% | |
| 3,4-Diaminobenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only