missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminothiophene dihydrochloride, 96%

CAS: 90069-81-1 | C4H6N22HCl | 187.09 g/mol

$139.35 - $139.35

Chemical Identifiers

CAS 90069-81-1
Molecular Formula C4H6N22HCl
Molecular Weight (g/mol) 187.09
MDL Number MFCD00671511
InChI Key RAMOMCXNLLLICQ-UHFFFAOYSA-N
Synonym 3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2
PubChem CID 12707342
IUPAC Name thiophene-3,4-diamine;dihydrochloride
SMILES C1=C(C(=CS1)N)N.Cl.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC325890010
SDS View Documents
Thermo Scientific Chemicals
325890010
1 g Glass bottle
Each for $139.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90069-81-1
187.09
RAMOMCXNLLLICQ-UHFFFAOYSA-N
12707342
C1=C(C(=CS1)N)N.Cl.Cl
C4H6N22HCl
MFCD00671511
3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2
thiophene-3,4-diamine;dihydrochloride

Specifications

90069-81-1
100.0
Brown or Gray
Authentic
Glass bottle
MFCD00671511
3,4-diaminothiophene dihydrochloride, thiophene-3,4-diamine dihydrochloride, 3,4-diaminothiophenedihydrochloride, pubchem21822, acmc-209r4q, 3,4-diaminothiophene 2hcl, 3?4-diaminothiophene dihydrochloride, 3,4-thiophenediamine, dihydrochloride, thiophene-3,4-diamine-hydrogen chloride 1/2
C1=C(C(=CS1)N)N.Cl.Cl
187.09
187.09
Crystalline Powder
95.0
240.0°C to 245.0°C
3% max.
95% min. (on dry substance) (Argentometry)
C4H6N22HCl
1 g
RAMOMCXNLLLICQ-UHFFFAOYSA-N
thiophene-3,4-diamine;dihydrochloride
12707342
96%
3, 4-Diaminothiophene dihydrochloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only