missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorophenethylamine, 99%, Thermo Scientific Chemicals

$249.50 - $249.50

Chemical Identifiers

CAS 21581-45-3
Molecular Formula C8H10Cl2N
Molecular Weight (g/mol) 191.07
MDL Number MFCD00060617
InChI Key MQPUAVYKVIHUJP-UHFFFAOYSA-O
Synonym 3,4-dichlorophenethylamine, 2-3,4-dichlorophenyl ethanamine, 2-3,4-dichlorophenyl ethylamine, 2-3,4-dichlorophenyl ethan-1-amine, 2-3,4-dichloro-phenyl-ethylamine, benzeneethanamine, 3,4-dichloro, 2-3,4,-dichlorophenyl ethylamine, 1-amino-2-3,4-dichlorophenyl ethane, pubchem12521, acmc-1apis
PubChem CID 217958
IUPAC Name 2-(3,4-dichlorophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=C(Cl)C(Cl)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350800050
SDS View Documents
Thermo Scientific Chemicals
350800050
5 g Glass bottle
Each for $249.50
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21581-45-3
191.07
MQPUAVYKVIHUJP-UHFFFAOYSA-O
217958
[NH3+]CCC1=CC=C(Cl)C(Cl)=C1
C8H10Cl2N
MFCD00060617
3,4-dichlorophenethylamine, 2-3,4-dichlorophenyl ethanamine, 2-3,4-dichlorophenyl ethylamine, 2-3,4-dichlorophenyl ethan-1-amine, 2-3,4-dichloro-phenyl-ethylamine, benzeneethanamine, 3,4-dichloro, 2-3,4,-dichlorophenyl ethylamine, 1-amino-2-3,4-dichlorophenyl ethane, pubchem12521, acmc-1apis
2-(3,4-dichlorophenyl)ethan-1-aminium

Specifications

21581-45-3
100.0
1.2680g/mL
98.5% min. (GC)
C8H10Cl2N
MFCD00060617
12, IV, 2491
3,4-dichlorophenethylamine, 2-3,4-dichlorophenyl ethanamine, 2-3,4-dichlorophenyl ethylamine, 2-3,4-dichlorophenyl ethan-1-amine, 2-3,4-dichloro-phenyl-ethylamine, benzeneethanamine, 3,4-dichloro, 2-3,4,-dichlorophenyl ethylamine, 1-amino-2-3,4-dichlorophenyl ethane, pubchem12521, acmc-1apis
[NH3+]CCC1=CC=C(Cl)C(Cl)=C1
191.07
190.07
Liquid
98.5
Colorless to Yellow
Authentic
Glass bottle
1.5655 to 1.5675
5 g
1.268
MQPUAVYKVIHUJP-UHFFFAOYSA-O
2-(3,4-dichlorophenyl)ethan-1-aminium
217958
99%
3, 4-Dichlorophenethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only