Learn More
3,4-Dichlorophenyl isocyanate, 97%, Thermo Scientific Chemicals
$78.30 - $248.85
Chemical Identifiers
CAS | 102-36-3 |
---|---|
Molecular Formula | C7H3Cl2NO |
Molecular Weight (g/mol) | 188.007 |
MDL Number | MFCD00002017 |
InChI Key | MFUVCHZWGSJKEQ-UHFFFAOYSA-N |
Synonym | 3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du |
PubChem CID | 7607 |
IUPAC Name | 1,2-dichloro-4-isocyanatobenzene |
SMILES | C1=CC(=C(C=C1N=C=O)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1629806
|
Thermo Scientific Chemicals
A1629806 |
5 g |
Each for $78.30
|
|
|||||
AAA1629814
|
Thermo Scientific Chemicals
A1629814 |
25 g |
Each for $248.85
|
|
|||||
Description
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C 'labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C ′″labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate
Solubility
Hydrolyzes in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, amines, alcohols.
Chemical Identifiers
102-36-3 | |
188.007 | |
MFUVCHZWGSJKEQ-UHFFFAOYSA-N | |
7607 | |
C1=CC(=C(C=C1N=C=O)Cl)Cl |
C7H3Cl2NO | |
MFCD00002017 | |
3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du | |
1,2-dichloro-4-isocyanatobenzene |
Specifications
102-36-3 | |
118°C to 120°C (18 mmHg) | |
Irritating | |
MFCD00002017 | |
UN2250 | |
Moisture sensitive | |
Hydrolyzes in water. | |
C1=CC(=C(C=C1N=C=O)Cl)Cl | |
188.007 | |
188.01 | |
3,4-Dichlorophenyl isocyanate |
40°C to 44°C | |
123°C (253°F) | |
C7H3Cl2NO | |
5 g | |
608325 | |
3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du | |
MFUVCHZWGSJKEQ-UHFFFAOYSA-N | |
1,2-dichloro-4-isocyanatobenzene | |
7607 | |
97% |
Safety and Handling
GHS H Statement
H300-H331-H334-H315-H319-H335
Fatal if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
H301-H312+H332-H315-H317-H319-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES
EINECSNumber : 203-026-2
RTECSNumber : NQ8760000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only