missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Diethoxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™

$154.05 - $620.14

Chemical Identifiers

CAS 5231-87-8
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
InChI Key DFSFLZCLKYZYRD-UHFFFAOYSA-N
Synonym 3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn
PubChem CID 123228
IUPAC Name 3,4-diethoxycyclobut-3-ene-1,2-dione
SMILES CCOC1=C(C(=O)C1=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D32535G
View Documents
TCI America
D32535G
5 g
Each for $154.05
 
D325325G
View Documents
TCI America
D325325G
25 g
Each for $620.14
 
Description

Description

Specifications

Chemical Identifiers

5231-87-8
170.164
3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn
3,4-diethoxycyclobut-3-ene-1,2-dione
C8H10O4
DFSFLZCLKYZYRD-UHFFFAOYSA-N
123228
CCOC1=C(C(=O)C1=O)OCC

Specifications

5231-87-8
95°C
5 g
DFSFLZCLKYZYRD-UHFFFAOYSA-N
3,4-diethoxycyclobut-3-ene-1,2-dione
123228
≥98.0% (GC)
3,4-Diethoxy-3-cyclobutene-1,2-dione
Yellow
C8H10O4
3,4-diethoxy-3-cyclobutene-1,2-dione, diethyl squarate, squaric acid diethyl ester, 3-cyclobutene-1,2-dione, 3,4-diethoxy, diethoxycyclobutenedione, diethoxycyclobut-3-ene-1,2-dione, 1,2-diethoxycyclobutenedione, 3,4-diethoxy-cyclobut-3-ene-1,2-dione, pubchem9741, acmc-209kyn
CCOC1=C(C(=O)C1=O)OCC
170.164
170.16
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : GU1772466

TSCA : No

SDS
Documents

Documents