missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl, 97%, Thermo Scientific™

$104.12 - $408.73

Chemical Identifiers

CAS 134412-18-3
Molecular Formula C20H22F2
Molecular Weight (g/mol) 300.39
MDL Number MFCD12828121
InChI Key TYZNCUASDJPXMP-UHFFFAOYSA-N
Synonym 3,4-difluoro-4'-4-ethylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-ethylcyclohexyl biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro-1,1'-biphenyl, 3,4-difluoro-4/'-4-ethylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-ethylcyclohexyl benzene, 4-4-4-ethylcyclohexyl phenyl-1,2-difluorobenzene, 1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro, 4'-4alpha-ethylcyclohexane-1beta-yl-3,4-difluorobiphenyl, 4-4-ethylcyclohexyl-3',4'-difluoro-1,1'-biphenyl #
PubChem CID 612308
SMILES CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5527503
View Documents
Thermo Scientific Chemicals
H5527503
1 g
Each for $104.12
 
AAH5527506
View Documents
Thermo Scientific Chemicals
H5527506
5 g
Each for $408.73
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

134412-18-3
300.39
TYZNCUASDJPXMP-UHFFFAOYSA-N
612308
C20H22F2
MFCD12828121
3,4-difluoro-4'-4-ethylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-ethylcyclohexyl biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro-1,1'-biphenyl, 3,4-difluoro-4/'-4-ethylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-ethylcyclohexyl benzene, 4-4-4-ethylcyclohexyl phenyl-1,2-difluorobenzene, 1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro, 4'-4alpha-ethylcyclohexane-1beta-yl-3,4-difluorobiphenyl, 4-4-ethylcyclohexyl-3',4'-difluoro-1,1'-biphenyl #
CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

134412-18-3
C20H22F2
1 g
TYZNCUASDJPXMP-UHFFFAOYSA-N
4'-(4-ethylcyclohexyl)-3,4-difluoro-1,1'-biphenyl
612308
97%
68°C to 69°C
MFCD12828121
3,4-difluoro-4'-4-ethylcyclohexyl biphenyl, 3,4-difluoro-4'-trans-4-ethylcyclohexyl biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro-1,1'-biphenyl, 3,4-difluoro-4/'-4-ethylcyclohexyl biphenyl, 1-3,4-difluorophenyl-4-4-ethylcyclohexyl benzene, 4-4-4-ethylcyclohexyl phenyl-1,2-difluorobenzene, 1,1'-biphenyl, 4'-4-ethylcyclohexyl-3,4-difluoro, 4'-4alpha-ethylcyclohexane-1beta-yl-3,4-difluorobiphenyl, 4-4-ethylcyclohexyl-3',4'-difluoro-1,1'-biphenyl #
CCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1
300.39
300.39
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'