missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™
$221.46 - $664.37
Chemical Identifiers
CAS | 85312-59-0 |
---|---|
Molecular Formula | C21H24F2 |
Molecular Weight (g/mol) | 314.42 |
MDL Number | MFCD09838999 |
InChI Key | VULXHDGYVHCLLN-UHFFFAOYSA-N |
Synonym | 4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl |
PubChem CID | 612306 |
IUPAC Name | 3,4-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl |
SMILES | CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 |
Chemical Identifiers
85312-59-0 | |
314.42 | |
VULXHDGYVHCLLN-UHFFFAOYSA-N | |
612306 | |
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 |
C21H24F2 | |
MFCD09838999 | |
4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl | |
3,4-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl |
Specifications
85312-59-0 | |
White | |
MFCD09838999 | |
4'-trans-4-propylcyclohexyl-3,4-difluorobiphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 1,1'-biphenyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl, 3,4-difluoro-4'-trans-4-propylcyclohexyl biphenyl, 1,2-difluoro-4-4-4-propylcyclohexyl phenyl benzene, 3,4-difluoro-4'-1s,4r-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl-1,1'-biphenyl, 3,4-difluoro-4'-4-propylcyclohexyl biphenyl, 4-4-propylcyclohexyl-3',4'-difluorobiphenyl, trans-3,4-difluoro-4'-4-n-propylcyclohexyl biphenyl | |
CCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C=C1 | |
314.42 | |
314.42 | |
Crystalline Powder |
68°C | |
C21H24F2 | |
5 g | |
VULXHDGYVHCLLN-UHFFFAOYSA-N | |
3,4-difluoro-4'-(4-propylcyclohexyl)-1,1'-biphenyl | |
612306 | |
≥98.0% (GC) | |
3,4-Difluoro-4′-(trans-4-propylcyclohexyl)biphenyl |
Safety and Handling
TSCA : No