missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorobenzenethiol 97.0+%, TCI America™
$259.11 - $1032.68
Chemical Identifiers
| CAS | 60811-24-7 |
|---|---|
| Molecular Formula | C6H3F2S |
| Molecular Weight (g/mol) | 145.15 |
| MDL Number | MFCD03094279 |
| InChI Key | BGVRHDQMTMPAEZ-UHFFFAOYSA-M |
| Synonym | 3,4-difluorothiophenol, 3,4-difluoro thiophenol, 3,4-difluorobenzene-1-thiol, benzenethiol, 3,4-difluoro, pubchem6772, 3,4-difluoro-thiophenol, 3.4-difluorobenzenethiol, acmc-1b3bf, 3,4-difluoro benzenethiol, 3,4-difluorobenzene-thiol |
| PubChem CID | 2778378 |
| IUPAC Name | (3,4-difluorophenyl)sulfanide |
| SMILES | FC1=CC=C([S-])C=C1F |
Chemical Identifiers
| 60811-24-7 | |
| 145.15 | |
| BGVRHDQMTMPAEZ-UHFFFAOYSA-M | |
| 2778378 | |
| FC1=CC=C([S-])C=C1F |
| C6H3F2S | |
| MFCD03094279 | |
| 3,4-difluorothiophenol, 3,4-difluoro thiophenol, 3,4-difluorobenzene-1-thiol, benzenethiol, 3,4-difluoro, pubchem6772, 3,4-difluoro-thiophenol, 3.4-difluorobenzenethiol, acmc-1b3bf, 3,4-difluoro benzenethiol, 3,4-difluorobenzene-thiol | |
| (3,4-difluorophenyl)sulfanide |
Specifications
| 60811-24-7 | |
| 70°C | |
| MFCD03094279 | |
| 3336 | |
| BGVRHDQMTMPAEZ-UHFFFAOYSA-M | |
| (3,4-difluorophenyl)sulfanide | |
| 2778378 | |
| ≥97.0% (GC) | |
| 3,4-Difluorobenzenethiol |
| Yellow | |
| C6H3F2S | |
| 5 g | |
| 3,4-difluorothiophenol, 3,4-difluoro thiophenol, 3,4-difluorobenzene-1-thiol, benzenethiol, 3,4-difluoro, pubchem6772, 3,4-difluoro-thiophenol, 3.4-difluorobenzenethiol, acmc-1b3bf, 3,4-difluoro benzenethiol, 3,4-difluorobenzene-thiol | |
| FC1=CC=C([S-])C=C1F | |
| 145.15 | |
| 146.15 | |
| Liquid |
Safety and Handling
TSCA : No