Learn More
3,4-Difluorophenylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific Chemicals
$216.90 - $819.33
Chemical Identifiers
CAS | 79784-34-2 |
---|---|
Molecular Formula | C8H4F2O2 |
Molecular Weight (g/mol) | 170.12 |
MDL Number | MFCD01733155 |
InChI Key | VZRYIZGMQICGSF-UHFFFAOYSA-N |
Synonym | 3,4-difluorophenylglyoxal hydrate, 2-3,4-difluorophenyl-2-oxoacetaldehyde hydrate, pubchem12210, 3,4-difluorophenylglyoxal monohydrate, 3,4-difluorophenylglyoxalhydrate, benzeneacetaldehyde,3,4-difluoro-a-oxo, 2-3,4-difluorophenyl-2-oxoethanal, hydrate |
PubChem CID | 2782289 |
IUPAC Name | 2-(3,4-difluorophenyl)-2-oxoacetaldehyde;hydrate |
SMILES | FC1=CC=C(C=C1F)C(=O)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2027103
|
Thermo Scientific Chemicals
L2027103 |
1 g |
Each for $216.90
|
|
|||||
AAL2027106
|
Thermo Scientific Chemicals
L2027106 |
5 g |
Each for $819.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
79784-34-2 | |
170.12 | |
VZRYIZGMQICGSF-UHFFFAOYSA-N | |
2782289 | |
FC1=CC=C(C=C1F)C(=O)C=O |
C8H4F2O2 | |
MFCD01733155 | |
3,4-difluorophenylglyoxal hydrate, 2-3,4-difluorophenyl-2-oxoacetaldehyde hydrate, pubchem12210, 3,4-difluorophenylglyoxal monohydrate, 3,4-difluorophenylglyoxalhydrate, benzeneacetaldehyde,3,4-difluoro-a-oxo, 2-3,4-difluorophenyl-2-oxoethanal, hydrate | |
2-(3,4-difluorophenyl)-2-oxoacetaldehyde;hydrate |
Spécifications
79784-34-2 | |
C8H4F2O2 | |
1 g | |
3,4-difluorophenylglyoxal hydrate, 2-3,4-difluorophenyl-2-oxoacetaldehyde hydrate, pubchem12210, 3,4-difluorophenylglyoxal monohydrate, 3,4-difluorophenylglyoxalhydrate, benzeneacetaldehyde,3,4-difluoro-a-oxo, 2-3,4-difluorophenyl-2-oxoethanal, hydrate | |
FC1=CC=C(C=C1F)C(=O)C=O | |
170.12 | |
170.12 (Anhydrous) | |
3,4-Difluorophenylglyoxal hydrate |
85°C to 90°C | |
MFCD01733155 | |
2574671 | |
VZRYIZGMQICGSF-UHFFFAOYSA-N | |
2-(3,4-difluorophenyl)-2-oxoacetaldehyde;hydrate | |
2782289 | |
98% | |
(dry wt.) |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'rtecsNumber' : CY1430000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only