missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13201CB
Specifications
3, 4-Dihydro-2H-1, 5-benzodioxepine-7-carboxylic acid | |
96.5 | |
97% | |
C10H10O4 | |
250mg | |
MQSSBVLREFSMDP-UHFFFAOYSA-N | |
3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid | |
2776383 | |
97% |
20825-89-2 | |
100.0 | |
Amber Glass Bottle | |
MFCD00276310 | |
3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro | |
C1COC2=C(C=C(C=C2)C(=O)O)OC1 | |
194.186 | |
194.19 |
Chemical Identifiers
20825-89-2 | |
194.186 | |
MQSSBVLREFSMDP-UHFFFAOYSA-N | |
2776383 | |
C1COC2=C(C=C(C=C2)C(=O)O)OC1 |
C10H10O4 | |
MFCD00276310 | |
3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carboxylic acid, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxylic acid, 2h-1,5-benzodioxepin-7-carboxylic acid, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-carboxylic acid, acmc-1crio, 7-carboxy-3,4-dihydro-2h-1,5-benzodioxepine, 2h-1,5-benzodioxepin-7-carboxylicacid, 3,4-dihydro, 2h-1,5-benzodioxepin-7-carboxylicacid,3,4-dihydro | |
3,4-dihydro-2H-1,5-benzodioxepine-7-carboxylic acid |