missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dihydro-2H-chromen-3-ylmethanol, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 76727-28-1 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.204 |
MDL Number | MFCD04972641 |
InChI Key | UPDSWAQIQZFOLF-UHFFFAOYSA-N |
Synonym | chroman-3-ylmethanol, 3,4-dihydro-2h-1-benzopyran-3-ylmethanol, 3-hydroxymethylchroman, chroman-3-yl-methanol, chroman-3-ylmethan-1-ol, 3,4-dihydro-2h-1-benzopyran 3-methanol, 2h-1-benzopyran-3-methanol,3,4-dihydro, 2h-1-benzopyran-3-methanol, 3,4-dihydro |
PubChem CID | 2795460 |
IUPAC Name | 3,4-dihydro-2H-chromen-3-ylmethanol |
SMILES | C1C(COC2=CC=CC=C21)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC25809CB
|
Thermo Scientific Chemicals
CC25809CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC25809DA
|
Thermo Scientific Chemicals
CC25809DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC25809DE
|
Thermo Scientific Chemicals
CC25809DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
76727-28-1 | |
164.204 | |
UPDSWAQIQZFOLF-UHFFFAOYSA-N | |
2795460 | |
C1C(COC2=CC=CC=C21)CO |
C10H12O2 | |
MFCD04972641 | |
chroman-3-ylmethanol, 3,4-dihydro-2h-1-benzopyran-3-ylmethanol, 3-hydroxymethylchroman, chroman-3-yl-methanol, chroman-3-ylmethan-1-ol, 3,4-dihydro-2h-1-benzopyran 3-methanol, 2h-1-benzopyran-3-methanol,3,4-dihydro, 2h-1-benzopyran-3-methanol, 3,4-dihydro | |
3,4-dihydro-2H-chromen-3-ylmethanol |
Specifications
76727-28-1 | |
100.0 | |
Amber Glass Bottle | |
MFCD04972641 | |
chroman-3-ylmethanol, 3,4-dihydro-2h-1-benzopyran-3-ylmethanol, 3-hydroxymethylchroman, chroman-3-yl-methanol, chroman-3-ylmethan-1-ol, 3,4-dihydro-2h-1-benzopyran 3-methanol, 2h-1-benzopyran-3-methanol,3,4-dihydro, 2h-1-benzopyran-3-methanol, 3,4-dihydro | |
C1C(COC2=CC=CC=C21)CO | |
164.204 | |
164.2 | |
3, 4-Dihydro-2H-chromen-3-ylmethanol |
96.5 | |
97% | |
C10H12O2 | |
250mg | |
UPDSWAQIQZFOLF-UHFFFAOYSA-N | |
3,4-dihydro-2H-chromen-3-ylmethanol | |
2795460 | |
97% |