missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 3,4-Dihydroxy-L-phenylalanine, 98+%

3-(3,4-Dihydroxyphenyl)-L-alanine (levodopa orL-dopa), CAS # 59-92-7, is a dopamine precursor molecule.

Supplier:  Thermo Scientific Chemicals A1131106

 View more versions of this product

Catalog No. AAA1131106


Explore available promotions

Description

Description

3,4-dihydroxy-L-phenylalanine (DOPA) is used as a cell adhesion molecule in serum-free cultures of anchorage-dependent mammalian cells. It is also used as an anticholinergic drug and used in the treatment of Parkinson's disease. It acts as an intermediate in the biosynthesis of other catecholamines (norepinephrine, epinephrine) and melanin. It acts as a precursor of dopamine product of tyrosine hydroxylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body
• Upon conversion to dopamine, it acts as an excitatory neurotransmitter
Applications
• L-dopa is useful in the laboratory study of Parkinson’s to replenish dopamine in depleted circuits
• L-dopa has also been used for laboratory research on hypertension and melanocyte activity
Specifications

Specifications

∼295°C (decomposition)
59-92-7
MFCD00002598
14,5464
Slightly soluble in water,dilute hydrochloric acid and formic acid. Insoluble in ethanol.
N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
197.19
6047
Odorless
≥98%
3,4-Dihydroxy-L-phenylalanine
C9H11NO4
2215169
levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
WTDRDQBEARUVNC-LURJTMIESA-N
(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
5 g
CHEBI:15765
197.19
Light sensitive

Chemical Identifiers

59-92-7
197.19
WTDRDQBEARUVNC-LURJTMIESA-N
6047
(2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
C9H11NO4
MFCD00002598
levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
CHEBI:15765
N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 200-445-2

RTECSNumber : AY5600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only