missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dihydroxyphenylacetic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1589314

 View more versions of this product

Catalog No. AAA1589314


Explore available promotions

Description

Description

3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxyphenylacetic acid is an important metabolite when studying the behavior of the dopaminergic system.

Solubility
Soluble in water and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air sensitive. Incompatible with strong bases and strong oxidizing agents. Store at room temperature
Specifications

Specifications

3,4-Dihydroxyphenylacetic acid
128°C to 130°C
MFCD00004338
2211017
Soluble in water and methanol.
C1=CC(=C(C=C1CC(=O)O)O)O
168.148
CHEBI:41941
≥98%
102-32-9
C8H8O4
25 g
3,4-dihydroxyphenylacetic acid, dopac, homoprotocatechuic acid, 2-3,4-dihydroxyphenyl acetic acid, dopacetic acid, benzeneacetic acid, 3,4-dihydroxy, dihydroxyphenylacetic acid, 3,4-dihydroxybenzeneacetic acid, 4-carboxymethylcatechol, homoprotocatechuate
CFFZDZCDUFSOFZ-UHFFFAOYSA-N
2-(3,4-dihydroxyphenyl)acetic acid
547
168.15

Chemical Identifiers

102-32-9
168.148
CFFZDZCDUFSOFZ-UHFFFAOYSA-N
547
2-(3,4-dihydroxyphenyl)acetic acid
C8H8O4
MFCD00004338
3,4-dihydroxyphenylacetic acid, dopac, homoprotocatechuic acid, 2-3,4-dihydroxyphenyl acetic acid, dopacetic acid, benzeneacetic acid, 3,4-dihydroxy, dihydroxyphenylacetic acid, 3,4-dihydroxybenzeneacetic acid, 4-carboxymethylcatechol, homoprotocatechuate
CHEBI:41941
C1=CC(=C(C=C1CC(=O)O)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 203-024-1

RTECSNumber : AH0590000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only