missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dimethoxy-L-phenylalanine, 97%, Thermo Scientific Chemicals
$156.14 - $514.99
Chemical Identifiers
CAS | 32161-30-1 |
---|---|
Molecular Formula | C11H15NO4 |
Molecular Weight (g/mol) | 225.244 |
MDL Number | MFCD01321274 |
InChI Key | VWTFNYVAFGYEKI-QMMMGPOBSA-N |
Synonym | 3,4-dimethoxy-l-phenylalanine, s-2-amino-3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl-l-alanine, l-3,4-dimethoxyphenylalanine, l-3,4-dimethoxyphenyl alanine, tyrosine, 3-methoxy-o-methyl, 3,4-dimethoxyphenylalanine, l, unii-gc6uei64ec, s-2-amino-3-3,4-dimethoxy-phenyl-propionic acid, l-tyrosine, 3-methoxy-o-methyl |
PubChem CID | 10632946 |
IUPAC Name | (2S)-2-amino-3-(3,4-dimethoxyphenyl)propanoic acid |
SMILES | COC1=C(C=C(C=C1)CC(C(=O)O)N)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5336803
|
Thermo Scientific Chemicals
H5336803 |
1 g |
Each for $156.14
|
|
|||||
AAH5336806
|
Thermo Scientific Chemicals
H5336806 |
5 g |
Each for $514.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32161-30-1 | |
225.244 | |
VWTFNYVAFGYEKI-QMMMGPOBSA-N | |
10632946 | |
COC1=C(C=C(C=C1)CC(C(=O)O)N)OC |
C11H15NO4 | |
MFCD01321274 | |
3,4-dimethoxy-l-phenylalanine, s-2-amino-3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl-l-alanine, l-3,4-dimethoxyphenylalanine, l-3,4-dimethoxyphenyl alanine, tyrosine, 3-methoxy-o-methyl, 3,4-dimethoxyphenylalanine, l, unii-gc6uei64ec, s-2-amino-3-3,4-dimethoxy-phenyl-propionic acid, l-tyrosine, 3-methoxy-o-methyl | |
(2S)-2-amino-3-(3,4-dimethoxyphenyl)propanoic acid |
Specifications
254°C to 257°C | |
C11H15NO4 | |
3,4-dimethoxy-l-phenylalanine, s-2-amino-3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl-l-alanine, l-3,4-dimethoxyphenylalanine, l-3,4-dimethoxyphenyl alanine, tyrosine, 3-methoxy-o-methyl, 3,4-dimethoxyphenylalanine, l, unii-gc6uei64ec, s-2-amino-3-3,4-dimethoxy-phenyl-propionic acid, l-tyrosine, 3-methoxy-o-methyl | |
COC1=C(C=C(C=C1)CC(C(=O)O)N)OC | |
1 g | |
10632946 | |
225.24 | |
97% |
32161-30-1 | |
MFCD01321274 | |
VWTFNYVAFGYEKI-QMMMGPOBSA-N | |
(2S)-2-amino-3-(3,4-dimethoxyphenyl)propanoic acid | |
225.244 | |
−5° (c=4 in 1M Hydrochloric Acid) | |
Air Sensitive | |
3,4-Dimethoxy-L-phenylalanine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only