missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethoxybenzylamine, 97%, Thermo Scientific Chemicals

$153.23 - $1957.66

Chemical Identifiers

CAS 5763-61-1
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00008116
InChI Key DIVNUTGTTIRPQA-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
PubChem CID 79832
IUPAC Name (3,4-dimethoxyphenyl)methanamine
SMILES COC1=CC=C(CN)C=C1OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2503306
View Documents
Thermo Scientific Chemicals
B2503306
5 g
Each for $153.23
 
AAB2503322
View Documents
Thermo Scientific Chemicals
B2503322
100 g
Each for $1,957.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5763-61-1
167.21
DIVNUTGTTIRPQA-UHFFFAOYSA-N
79832
COC1=CC=C(CN)C=C1OC
C9H13NO2
MFCD00008116
3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
(3,4-dimethoxyphenyl)methanamine

Specifications

5763-61-1
281°C to 284°C
C9H13NO2
MFCD00008116
UN2735
Air Sensitive
DIVNUTGTTIRPQA-UHFFFAOYSA-N
(3,4-dimethoxyphenyl)methanamine
79832
97%
1.109
>110°C (230°F)
1.556
5 g
511575
3,4-dimethoxybenzylamine, veratrylamine, 3,4-dimethoxyphenyl methanamine, benzenemethanamine, 3,4-dimethoxy, 1-3,4-dimethoxyphenyl methanamine, benzylamine, 3,4-dimethoxy, 3,4-dimethoxyphenyl methylamine, veratrylamin, pubchem7394, acmc-1altc
COC1=CC=C(CN)C=C1OC
167.21
167.21
3,4-Dimethoxybenzylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 227-287-7

RTECSNumber : YX7355000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only