missing translation for 'onlineSavingsMsg'
Learn More

(3,4-Dimethoxyphenyl)acetonitrile, 99+%

CAS: 93-17-4 | C10H11NO2 | 177.2 g/mol

$47.25 - $121.03

Chemical Identifiers

CAS 93-17-4
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00001911
InChI Key ASLSUMISAQDOOB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
PubChem CID 66727
IUPAC Name 2-(3,4-dimethoxyphenyl)acetonitrile
SMILES COC1=C(C=C(C=C1)CC#N)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC155260250
SDS View Documents
Thermo Scientific Chemicals
155260250
25 g Glass bottle
Each for $47.25
Only null left
 
AC155261000
SDS View Documents
Thermo Scientific Chemicals
155261000
100 g Glass bottle
Each for $121.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

93-17-4
177.2
ASLSUMISAQDOOB-UHFFFAOYSA-N
66727
COC1=C(C=C(C=C1)CC#N)OC
C10H11NO2
MFCD00001911
3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
2-(3,4-dimethoxyphenyl)acetonitrile

Specifications

93-17-4
100.0
White
250°C
Authentic
Glass bottle
(CH3O)2C6H3CH2CN
25 g
3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
ASLSUMISAQDOOB-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)acetonitrile
66727
99+%
(3, 4-Dimethoxyphenyl)acetonitrile
99.0
62°C to 65°C
171°C to 178°C (10.0 mmHg)
0.5% max. (vacuum)
99+%
C10H11NO2
MFCD00001911
10, I, 198
Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,chloroform and,ethanol
COC1=C(C=C(C=C1)CC#N)OC
177.2
177.2
Crystals or Crystalline Needles
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-225-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only