Learn More
3,4-Dimethoxyphenylmagnesium bromide, 0.5M in THF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H54654AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Air and moisture sensitive. Air, oxidizing agents, heat are incompatible. Store under cool place.
Specifications
3,4-Dimethoxyphenylmagnesium bromide | |
C8H9BrMgO2 | |
100 mL | |
Air and moisture sensitive; May form a precipitate | |
Reacts with water. | |
COC1=CC=C([Mg]Br)C=C1OC | |
241.37 | |
241.37 |
89980-69-8 | |
MFCD01311476 | |
UN2924 | |
3,4-dimethoxyphenylmagnesium bromide, 3,4-dimethoxyphenyl magnesium bromide, 3,4-dimethoxyphenylmagnesium bromide solution, bhtpzwcaihfrsy-uhfffaoysa-m, 3,4-dimethoxyphenylmagnesium bromide, 0.5m in thf, 3,4-dimethoxyphenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,4-dimethoxyphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent | |
BHTPZWCAIHFRSY-UHFFFAOYSA-M | |
magnesium;1,2-dimethoxybenzene-5-ide;bromide | |
11010104 | |
0.5M in THF |
Chemical Identifiers
89980-69-8 | |
241.37 | |
BHTPZWCAIHFRSY-UHFFFAOYSA-M | |
11010104 | |
COC1=CC=C([Mg]Br)C=C1OC |
C8H9BrMgO2 | |
MFCD01311476 | |
3,4-dimethoxyphenylmagnesium bromide, 3,4-dimethoxyphenyl magnesium bromide, 3,4-dimethoxyphenylmagnesium bromide solution, bhtpzwcaihfrsy-uhfffaoysa-m, 3,4-dimethoxyphenylmagnesium bromide, 0.5m in thf, 3,4-dimethoxyphenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,4-dimethoxyphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent | |
magnesium;1,2-dimethoxybenzene-5-ide;bromide |
Safety and Handling
GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280i-P281-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P363-P370+P378q-P5
H225-H300+H310+H330-H319-H335-H336-H351
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only