Learn More
3,4-Dimethyl-1-pentyn-3-ol, 94%, Thermo Scientific Chemicals
$70.01 - $252.21
Chemical Identifiers
CAS | 1482-15-1 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.17 |
MDL Number | MFCD00039845 |
InChI Key | DZNLEQBXXLGELU-UHFFFAOYNA-N |
PubChem CID | 95483 |
IUPAC Name | 3,4-dimethylpent-1-yn-3-ol |
SMILES | CC(C)C(C)(O)C#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1990606
|
Thermo Scientific Chemicals
A1990606 |
5 g |
Each for $70.01
|
|
|||||
AAA1990614
|
Thermo Scientific Chemicals
A1990614 |
25 g |
Each for $252.21
|
|
|||||
Description
3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.
Solubility
Fully miscible in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Chemical Identifiers
1482-15-1 | |
112.17 | |
DZNLEQBXXLGELU-UHFFFAOYNA-N | |
3,4-dimethylpent-1-yn-3-ol |
C7H12O | |
MFCD00039845 | |
95483 | |
CC(C)C(C)(O)C#C |
Specifications
1482-15-1 | |
133°C | |
C7H12O | |
5 g | |
3,4-dimethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3,4-dimethyl, acmc-1bzc3, 2-isopropyl-3-butyne-2-ol, 1-pentyn-3-ol,4-dimethyl, 3,4-dimethyl-pent-1-yn-3-ol, dznleqbxxlgelu-uhfffaoysa, 1-pentyn-3-ol,3,4-dimethyl, 1,2-dimethyl-1-hydroxypropyl acetylene | |
DZNLEQBXXLGELU-UHFFFAOYNA-N | |
3,4-dimethylpent-1-yn-3-ol | |
95483 | |
94% |
0.87 | |
40°C (104°F) | |
MFCD00039845 | |
UN1987 | |
Fully miscible in water. | |
CC(C)C(C)(O)C#C | |
112.17 | |
112.17 | |
3,4-Dimethyl-1-pentyn-3-ol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only