missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5973-71-7
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00016612
InChI Key POQJHLBMLVTHAU-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
PubChem CID 22278
IUPAC Name 3,4-dimethylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC346060050
SDS View Documents
Thermo Scientific Chemicals
346060050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC346060250
SDS View Documents
Thermo Scientific Chemicals
346060250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5973-71-7
134.18
POQJHLBMLVTHAU-UHFFFAOYSA-N
22278
CC1=C(C=C(C=C1)C=O)C
C9H10O
MFCD00016612
benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
3,4-dimethylbenzaldehyde

Specifications

5973-71-7
100.0
226°C
96% min. (GC)
C9H10O
(CH3)2C6H3CHO
5g
1.012
POQJHLBMLVTHAU-UHFFFAOYSA-N
3,4-dimethylbenzaldehyde
22278
97%
96.0
1.0120g/mL
Authentic
Glass bottle
1.5490 to 1.551
MFCD00016612
07,IV,707
benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
CC1=C(C=C(C=C1)C=O)C
134.18
134.18
3, 4-Dimethylbenzaldehyde, 98%
Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

EINECSNumber : 227-770-2

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only