missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzoic Acid 99.0+%, TCI America™

$80.16 - $134.21

Chemical Identifiers

CAS 619-04-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002524
InChI Key OPVAJFQBSDUNQA-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dimethyl, 3,4-dimethyl-benzoic acid, 1-carboxy-3,4-dimethylbenzene, o-xylene-4-carboxylic acid, 3,4-dimethylbenzoicacid, pubchem14893, 3,4-dimethyl benzoic acid, asym-o-xylylic acid, acmc-1b5p1, 3, 4 dimethyl benzoic acid
PubChem CID 12073
ChEBI CHEBI:64818
IUPAC Name 3,4-dimethylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D143910G
View Documents
TCI America
D143910G
10 g
Each for $80.16
Only null left
 
D143925G
View Documents
TCI America
D143925G
25 g
Each for $134.21
Only null left
 
Description

Description

Specifications

Chemical Identifiers

619-04-5
150.177
OPVAJFQBSDUNQA-UHFFFAOYSA-N
12073
3,4-dimethylbenzoic acid
C9H10O2
MFCD00002524
benzoic acid, 3,4-dimethyl, 3,4-dimethyl-benzoic acid, 1-carboxy-3,4-dimethylbenzene, o-xylene-4-carboxylic acid, 3,4-dimethylbenzoicacid, pubchem14893, 3,4-dimethyl benzoic acid, asym-o-xylylic acid, acmc-1b5p1, 3, 4 dimethyl benzoic acid
CHEBI:64818
CC1=C(C=C(C=C1)C(=O)O)C

Specifications

166°C
White-Yellow
Crystal-Powder at 20°C
MFCD00002524
OPVAJFQBSDUNQA-UHFFFAOYSA-N
3,4-dimethylbenzoic acid
12073
150.18
3,4-Dimethylbenzoic Acid
619-04-5
10 g
C9H10O2
benzoic acid, 3,4-dimethyl, 3,4-dimethyl-benzoic acid, 1-carboxy-3,4-dimethylbenzene, o-xylene-4-carboxylic acid, 3,4-dimethylbenzoicacid, pubchem14893, 3,4-dimethyl benzoic acid, asym-o-xylylic acid, acmc-1b5p1, 3, 4 dimethyl benzoic acid
CC1=C(C=C(C=C1)C(=O)O)C
150.177
CHEBI:64818
≥99.0% (T)
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DG8734020

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'