Learn More
3,4-Dimethylhexane, 97%, Thermo Scientific Chemicals
$132.67 - $506.26
Chemical Identifiers
CAS | 583-48-2 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.232 |
MDL Number | MFCD00039927 |
InChI Key | RNTWWGNZUXGTAX-UHFFFAOYSA-N |
Synonym | hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso |
PubChem CID | 11412 |
IUPAC Name | 3,4-dimethylhexane |
SMILES | CCC(C)C(C)CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5349806
|
Thermo Scientific Chemicals
H5349806 |
5 g |
Each for $132.67
|
|
|||||
AAH5349814
|
Thermo Scientific Chemicals
H5349814 |
25 g |
Each for $506.26
|
|
|||||
Description
3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
583-48-2 | |
114.232 | |
RNTWWGNZUXGTAX-UHFFFAOYSA-N | |
11412 | |
CCC(C)C(C)CC |
C8H18 | |
MFCD00039927 | |
hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso | |
3,4-dimethylhexane |
Specifications
583-48-2 | |
119°C | |
C8H18 | |
MFCD00039927 | |
UN1262 | |
hexane, 3,4-dimethyl, 3,4-dimethyl hexane, acmc-1aquh, c2h5ch ch3 ch ch3 c2h5, 3,4-dimethylhexane, analytical standard, 3,4-dimethylhexane, mixture of +/-and meso | |
CCC(C)C(C)CC | |
114.232 | |
114.23 | |
3,4-Dimethylhexane |
0.72 | |
6°C (43°F) | |
1.403 | |
5 g | |
1718744 | |
RNTWWGNZUXGTAX-UHFFFAOYSA-N | |
3,4-dimethylhexane | |
11412 | |
97% |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECSNumber : 209-504-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only