missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylacetic acid, 98%

CAS: 17283-16-8 | C10H11O2 | 163.20 g/mol

$87.55 - $330.11

Chemical Identifiers

CAS 17283-16-8
Molecular Formula C10H11O2
Molecular Weight (g/mol) 163.20
MDL Number MFCD02664684
InChI Key OTTPBKINJOYJGW-UHFFFAOYSA-M
PubChem CID 296013
IUPAC Name 2-(3,4-dimethylphenyl)acetic acid
SMILES CC1=CC=C(CC([O-])=O)C=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6317803
View Documents
Thermo Scientific Chemicals
H6317803
1 g
Each for $87.55
Only null left
 
AAH6317806
View Documents
Thermo Scientific Chemicals
H6317806
5 g
Each for $330.11
Only null left
 
Description

Description

3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

17283-16-8
163.20
OTTPBKINJOYJGW-UHFFFAOYSA-M
2-(3,4-dimethylphenyl)acetic acid
C10H11O2
MFCD02664684
296013
CC1=CC=C(CC([O-])=O)C=C1C

Specifications

17283-16-8
Solid
MFCD02664684
Slightly soluble in water.
CC1=CC=C(CC([O-])=O)C=C1C
163.20
164.2
3,4-Dimethylphenylacetic acid
1 g
C10H11O2
2-3,4-dimethylphenyl acetic acid, 3,4-dimethylphenylacetic acid, 3,4-dimethyl-phenyl-acetic acid, 3,4-dimethylphenylaceticacid, 3,4-dimethyphenylacetic acid, 3,4-dimethylphenyl acetic acid, benzeneacetic acid, 3,4-dimethyl, 3,4-dimethylbenzenacetic acid, pubchem10440, benzeneacetic acid,3,4-dimethyl
OTTPBKINJOYJGW-UHFFFAOYSA-M
2-(3,4-dimethylphenyl)acetic acid
296013
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only