missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(4-hydroxyphenyl)propanohydrazide, Thermo Scientific™
Chemical Identifiers
CAS | 65330-63-4 |
---|---|
Molecular Formula | C9H12N2O2 |
Molecular Weight (g/mol) | 180.21 |
MDL Number | MFCD00025134 |
InChI Key | JMNHUOPRGLTNJI-UHFFFAOYSA-N |
Synonym | 3-4-hydroxyphenyl propanohydrazide, 3-4-hydroxyphenyl propanehydrazide, 3-4-hydroxyphenyl propionohydrazide, benzenepropanoic acid,4-hydroxy-, hydrazide, benzenepropanoic acid, 4-hydroxy-, hydrazide |
PubChem CID | 2796587 |
IUPAC Name | 3-(4-hydroxyphenyl)propanehydrazide |
SMILES | NNC(=O)CCC1=CC=C(O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
BTB00937DE
|
Thermo Scientific Chemicals
BTB00937DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB00937EA
|
Thermo Scientific Chemicals
BTB00937EA |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
65330-63-4 | |
180.21 | |
JMNHUOPRGLTNJI-UHFFFAOYSA-N | |
2796587 | |
NNC(=O)CCC1=CC=C(O)C=C1 |
C9H12N2O2 | |
MFCD00025134 | |
3-4-hydroxyphenyl propanohydrazide, 3-4-hydroxyphenyl propanehydrazide, 3-4-hydroxyphenyl propionohydrazide, benzenepropanoic acid,4-hydroxy-, hydrazide, benzenepropanoic acid, 4-hydroxy-, hydrazide | |
3-(4-hydroxyphenyl)propanehydrazide |
Specifications
65330-63-4 | |
C9H12N2O2 | |
5g | |
JMNHUOPRGLTNJI-UHFFFAOYSA-N | |
3-(4-hydroxyphenyl)propanehydrazide | |
2796587 | |
97% |
Amber glass bottle | |
MFCD00025134 | |
3-4-hydroxyphenyl propanohydrazide, 3-4-hydroxyphenyl propanehydrazide, 3-4-hydroxyphenyl propionohydrazide, benzenepropanoic acid,4-hydroxy-, hydrazide, benzenepropanoic acid, 4-hydroxy-, hydrazide | |
NNC(=O)CCC1=CC=C(O)C=C1 | |
180.21 | |
180.21 | |
3-(4-hydroxyphenyl)propanohydrazide |
Safety and Handling
GHS Signal Word: Warning