missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Hydroxyphenyl)propionic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 121751000

 View more versions of this product

Catalog No. AC121751000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-(4-Hydroxyphenyl)propionic acid
97.5
Authentic
Glass bottle
HOC6H4CH2CH2CO2H
100g
3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid
NMHMNPHRMNGLLB-UHFFFAOYSA-N
3-(4-hydroxyphenyl)propanoic acid
10394
166.18
501-97-3
100.0
98%
C9H10O3
MFCD00002778
10, 244
Solubility in water: slightly soluble in water. Other solubilities: soluble in alchol,acetone and ether
C1=CC(=CC=C1CCC(=O)O)O
166.18
CHEBI:32980
98%

Chemical Identifiers

501-97-3
166.18
NMHMNPHRMNGLLB-UHFFFAOYSA-N
10394
3-(4-hydroxyphenyl)propanoic acid
C9H10O3
MFCD00002778
3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid
CHEBI:32980
C1=CC(=CC=C1CCC(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-931-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only