missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Methyl-1-piperazinyl)aniline 98.0+%, TCI America™

$68.97 - $274.02

Chemical Identifiers

CAS 148546-99-0
Molecular Formula C11H17N3
Molecular Weight (g/mol) 191.278
MDL Number MFCD08435857
InChI Key RJGHJWKQCJAJEP-UHFFFAOYSA-N
Synonym 3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
PubChem CID 11564613
IUPAC Name 3-(4-methylpiperazin-1-yl)aniline
SMILES CN1CCN(CC1)C2=CC(=CC=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A27141G
View Documents
TCI America
A27141G
1 g N/A
Only null left
 
A27145G
View Documents
TCI America
A27145G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

148546-99-0
191.278
RJGHJWKQCJAJEP-UHFFFAOYSA-N
11564613
CN1CCN(CC1)C2=CC(=CC=C2)N
C11H17N3
MFCD08435857
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
3-(4-methylpiperazin-1-yl)aniline

Specifications

148546-99-0
C11H17N3
1 g
3-4-methylpiperazin-1-yl aniline, 3-4-methyl-1-piperazinyl aniline, 3-4-methylpiperazin-1-yl benzenamine, benzenamine, 3-4-methyl-1-piperazinyl, zlchem 128, acmc-20a0ig, ksc496g5r, 3-4-methyl-piperazin-1-ylaniline, 3-4-methylpiperazinyl phenylamine, 3-4-methylpiperazine-1-yl aniline
CN1CCN(CC1)C2=CC(=CC=C2)N
191.278
191.28
Crystalline Powder
85°C
MFCD08435857
3263
RJGHJWKQCJAJEP-UHFFFAOYSA-N
3-(4-methylpiperazin-1-yl)aniline
11564613
≥98.0% (GC,T)
3-(4-Methyl-1-piperazinyl)aniline
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'