missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Nitrophenyl)propionic acid, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3366318

 View more versions of this product

Catalog No. AAH3366318


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

3-(4-Nitrophenyl)propionic acid
164°C to 168°C
MFCD00126834
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
VZOPVJNBOQOLPN-UHFFFAOYSA-N
3-(4-nitrophenyl)propanoic acid
85526
96%
16642-79-8
C9H9NO4
50 g
Slightly soluble in water.
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O
195.17
195.17

Chemical Identifiers

16642-79-8
195.17
VZOPVJNBOQOLPN-UHFFFAOYSA-N
85526
OC(=O)CCC1=CC=C(C=C1)[N+]([O-])=O
C9H9NO4
MFCD00126834
3-4-nitrophenyl propanoic acid, 3-4-nitrophenyl propionic acid, benzenepropanoic acid, 4-nitro, 3-4-nitrophenyl-propionic acid, 4-nitrobenzenepropanoic acid, p-nitrohydrozimtsaure, pubchem13615, p-nitrohydrocinnamic acid
3-(4-nitrophenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only