missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(4-tert-Butylphenyl)isobutyraldehyde 96.0+%, TCI America™
$57.27 - $428.57
Chemical Identifiers
CAS | 80-54-6 |
---|---|
Molecular Formula | C14H20O |
Molecular Weight (g/mol) | 204.31 |
MDL Number | MFCD00047655 |
InChI Key | SDQFDHOLCGWZPU-UHFFFAOYNA-N |
Synonym | lilial, 3-4-tert-butyl phenyl-2-methylpropanal, 3-4-tert-butylphenyl isobutyraldehyde, lilyal, lily aldehyde, 3-4-tert-butylphenyl-2-methylpropanal, 3-4-tert-butylphenyl-2-methylpropionaldehyde, benzenepropanal, 4-1,1-dimethylethyl-.alpha.-methyl, butylphenyl methylpropional, 2-4-tert-butylbenzyl propionaldehyde |
PubChem CID | 228987 |
IUPAC Name | 3-(4-tert-butylphenyl)-2-methylpropanal |
SMILES | CC(CC1=CC=C(C=C1)C(C)(C)C)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
B114525ML
|
TCI America
B114525ML |
25 mL |
Each for $57.27
|
|
||||||
B1145100ML
|
TCI America
B1145100ML |
100 mL |
Each for $144.17
|
|
||||||
B1145500ML
|
TCI America
B1145500ML |
500 mL |
Each for $428.57
|
|
||||||
Chemical Identifiers
80-54-6 | |
204.31 | |
SDQFDHOLCGWZPU-UHFFFAOYNA-N | |
228987 | |
CC(CC1=CC=C(C=C1)C(C)(C)C)C=O |
C14H20O | |
MFCD00047655 | |
lilial, 3-4-tert-butyl phenyl-2-methylpropanal, 3-4-tert-butylphenyl isobutyraldehyde, lilyal, lily aldehyde, 3-4-tert-butylphenyl-2-methylpropanal, 3-4-tert-butylphenyl-2-methylpropionaldehyde, benzenepropanal, 4-1,1-dimethylethyl-.alpha.-methyl, butylphenyl methylpropional, 2-4-tert-butylbenzyl propionaldehyde | |
3-(4-tert-butylphenyl)-2-methylpropanal |
Specifications
80-54-6 | |
150°C | |
MFCD00047655 | |
lilial, 3-4-tert-butyl phenyl-2-methylpropanal, 3-4-tert-butylphenyl isobutyraldehyde, lilyal, lily aldehyde, 3-4-tert-butylphenyl-2-methylpropanal, 3-4-tert-butylphenyl-2-methylpropionaldehyde, benzenepropanal, 4-1,1-dimethylethyl-.alpha.-methyl, butylphenyl methylpropional, 2-4-tert-butylbenzyl propionaldehyde | |
CC(CC1=CC=C(C=C1)C(C)(C)C)C=O | |
204.31 | |
204.31 | |
Liquid |
Yellow | |
C14H20O | |
25 mL | |
SDQFDHOLCGWZPU-UHFFFAOYNA-N | |
3-(4-tert-butylphenyl)-2-methylpropanal | |
228987 | |
≥96.0% (GC) | |
3-(4-tert-Butylphenyl)isobutyraldehyde |
Safety and Handling
EINECSNumber : (3)-2667
RTECSNumber : MW4895000
TSCA : Yes