missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-tert-Butylphenyl)propionic acid, 97%, Thermo Scientific Chemicals

$81.81 - $940.48

Chemical Identifiers

CAS 1208-64-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00796482
InChI Key BNJYANVQFVSYEK-UHFFFAOYSA-N
Synonym 3-4-tert-butyl-phenyl-propionic acid, 3-4-tert-butyl phenyl propanoic acid, 3-4-tert-butylphenyl propanoic acid, 4-1,1-dimethylethyl benzenepropanoic acid, 4-t-butylhydrocinnamic acid, 4-t-butylhydrocinnamic aicd, 4-tert-butylhydrocinnamic acid, 3-4-t-butylphenyl propionic acid, 3-4-t-butylphenyl propanoic acid
PubChem CID 4962173
IUPAC Name 3-(4-tert-butylphenyl)propanoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)CCC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5401103
View Documents
Thermo Scientific Chemicals
H5401103
1 g
Each for $81.81
 
AAH5401106
View Documents
Thermo Scientific Chemicals
H5401106
5 g
Each for $228.27
 
AAH5401114
View Documents
Thermo Scientific Chemicals
H5401114
25 g
Each for $940.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1208-64-6
206.285
BNJYANVQFVSYEK-UHFFFAOYSA-N
4962173
CC(C)(C)C1=CC=C(C=C1)CCC(=O)O
C13H18O2
MFCD00796482
3-4-tert-butyl-phenyl-propionic acid, 3-4-tert-butyl phenyl propanoic acid, 3-4-tert-butylphenyl propanoic acid, 4-1,1-dimethylethyl benzenepropanoic acid, 4-t-butylhydrocinnamic acid, 4-t-butylhydrocinnamic aicd, 4-tert-butylhydrocinnamic acid, 3-4-t-butylphenyl propionic acid, 3-4-t-butylphenyl propanoic acid
3-(4-tert-butylphenyl)propanoic acid

Specifications

1208-64-6
C13H18O2
1 g
BNJYANVQFVSYEK-UHFFFAOYSA-N
3-(4-tert-butylphenyl)propanoic acid
4962173
97%
102°C
MFCD00796482
3-4-tert-butyl-phenyl-propionic acid, 3-4-tert-butyl phenyl propanoic acid, 3-4-tert-butylphenyl propanoic acid, 4-1,1-dimethylethyl benzenepropanoic acid, 4-t-butylhydrocinnamic acid, 4-t-butylhydrocinnamic aicd, 4-tert-butylhydrocinnamic acid, 3-4-t-butylphenyl propionic acid, 3-4-t-butylphenyl propanoic acid
CC(C)(C)C1=CC=C(C=C1)CCC(=O)O
206.285
206.28
3-(4-tert-Butylphenyl)propionic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only