missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5,5-Trimethylhexanoyl Chloride 90.0+%, TCI America™
Supplier: TCI America T095125ML
Specifications
3,5,5-Trimethylhexanoyl Chloride | |
Yellow | |
C9H17ClO | |
25 mL | |
GEKPNPPFAYJZRD-UHFFFAOYSA-N | |
3,5,5-trimethylhexanoyl chloride | |
161929 | |
≥90.0% (GC) |
36727-29-4 | |
190°C | |
MFCD00044028 | |
3265 | |
CC(CC(=O)Cl)CC(C)(C)C | |
176.684 | |
176.68 | |
Liquid |
Chemical Identifiers
36727-29-4 | |
176.684 | |
GEKPNPPFAYJZRD-UHFFFAOYSA-N | |
3,5,5-trimethylhexanoyl chloride |
C9H17ClO | |
MFCD00044028 | |
161929 | |
CC(CC(=O)Cl)CC(C)(C)C |
Safety and Handling
RTECSNumber : MP4560000
TSCA : Yes