missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Bis[3,5-bis(methoxycarbonyl)phenoxymethyl]phenol 98.0+%, TCI America™
Supplier: TCI America B2865200MG
Specifications
3,5-Bis[3,5-bis(methoxycarbonyl)phenoxymethyl]phenol | |
163°C | |
C28H26O11 | |
USUINPRIDFWVMZ-UHFFFAOYSA-N | |
dimethyl 5-[[3-[[3,5-bis(methoxycarbonyl)phenoxy]methyl]-5-hydroxyphenyl]methoxy]benzene-1,3-dicarboxylate | |
11124374 | |
≥98.0% (HPLC) |
186605-76-5 | |
White-Yellow | |
200 mg | |
COC(=O)C1=CC(=CC(=C1)OCC2=CC(=CC(=C2)O)COC3=CC(=CC(=C3)C(=O)OC)C(=O)OC)C(=O)OC | |
538.505 | |
538.51 | |
Crystalline Powder |
Chemical Identifiers
186605-76-5 | |
538.505 | |
11124374 | |
COC(=O)C1=CC(=CC(=C1)OCC2=CC(=CC(=C2)O)COC3=CC(=CC(=C3)C(=O)OC)C(=O)OC)C(=O)OC |
C28H26O11 | |
USUINPRIDFWVMZ-UHFFFAOYSA-N | |
dimethyl 5-[[3-[[3,5-bis(methoxycarbonyl)phenoxy]methyl]-5-hydroxyphenyl]methoxy]benzene-1,3-dicarboxylate |
Safety and Handling
TSCA : No