missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Bis(methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

$318.26 - $1617.51

Chemical Identifiers

CAS 177735-55-6
Molecular Formula C10H11BO6
Molecular Weight (g/mol) 238.00
MDL Number MFCD11053854
InChI Key WEJWFDLAZSVCJK-UHFFFAOYSA-N
Synonym 3,5-bis methoxycarbonyl phenylboronic acid, 3,5-bis methoxycarbonyl phenyl boronic acid, 3,5-bis methoxycarbonyl benzeneboronic acid, 1,3-benzenedicarboxylic acid, 5-borono-, 1,3-dimethyl ester, 3,5-di methoxycarbonyl phenylboronic acid, acmc-1c6yh, 3,5-bis methoxycarbonyl phenyl-dihydroxyborane
PubChem CID 19363104
IUPAC Name [3,5-bis(methoxycarbonyl)phenyl]boronic acid
SMILES COC(=O)C1=CC(=CC(=C1)B(O)O)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6447203
View Documents
Thermo Scientific Chemicals
H6447203
1 g
Each for $318.26
 
AAH6447206
View Documents
Thermo Scientific Chemicals
H6447206
5 g
Each for $1,617.51
 
Description

Description

It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the for the synthesis of 3,4-disubstituted coumarins via Pd-Catalyzed site-selective cross-coupling reactions.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

177735-55-6
238.00
WEJWFDLAZSVCJK-UHFFFAOYSA-N
19363104
COC(=O)C1=CC(=CC(=C1)B(O)O)C(=O)OC
C10H11BO6
MFCD11053854
3,5-bis methoxycarbonyl phenylboronic acid, 3,5-bis methoxycarbonyl phenyl boronic acid, 3,5-bis methoxycarbonyl benzeneboronic acid, 1,3-benzenedicarboxylic acid, 5-borono-, 1,3-dimethyl ester, 3,5-di methoxycarbonyl phenylboronic acid, acmc-1c6yh, 3,5-bis methoxycarbonyl phenyl-dihydroxyborane
[3,5-bis(methoxycarbonyl)phenyl]boronic acid

Specifications

177735-55-6
MFCD11053854
3,5-bis methoxycarbonyl phenylboronic acid, 3,5-bis methoxycarbonyl phenyl boronic acid, 3,5-bis methoxycarbonyl benzeneboronic acid, 1,3-benzenedicarboxylic acid, 5-borono-, 1,3-dimethyl ester, 3,5-di methoxycarbonyl phenylboronic acid, acmc-1c6yh, 3,5-bis methoxycarbonyl phenyl-dihydroxyborane
COC(=O)C1=CC(=CC(=C1)B(O)O)C(=O)OC
238.00
238
Solid
C10H11BO6
1 g
WEJWFDLAZSVCJK-UHFFFAOYSA-N
[3,5-bis(methoxycarbonyl)phenyl]boronic acid
19363104
97%
3,5-Bis(methoxycarbonyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only