missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(methoxymethyloxy)benzyl Alcohol 98.0+%, TCI America™
$234.87 - $700.89
Chemical Identifiers
| CAS | 76280-60-9 |
|---|---|
| Molecular Formula | C11H16O5 |
| Molecular Weight (g/mol) | 228.24 |
| MDL Number | MFCD27928465 |
| InChI Key | QZORVGYVDDOQNT-UHFFFAOYSA-N |
| Synonym | [3,5-Bis(methoxymethoxy)phenyl]methanol |
| PubChem CID | 10443751 |
| IUPAC Name | [3,5-bis(methoxymethoxy)phenyl]methanol |
| SMILES | COCOC1=CC(OCOC)=CC(CO)=C1 |
Chemical Identifiers
| 76280-60-9 | |
| 228.24 | |
| QZORVGYVDDOQNT-UHFFFAOYSA-N | |
| 10443751 | |
| COCOC1=CC(OCOC)=CC(CO)=C1 |
| C11H16O5 | |
| MFCD27928465 | |
| [3,5-Bis(methoxymethoxy)phenyl]methanol | |
| [3,5-bis(methoxymethoxy)phenyl]methanol |
Specifications
| 76280-60-9 | |
| 172°C | |
| MFCD27928465 | |
| [3,5-Bis(methoxymethoxy)phenyl]methanol | |
| COCOC1=CC(OCOC)=CC(CO)=C1 | |
| 228.24 | |
| 228.24 | |
| Liquid |
| Yellow | |
| C11H16O5 | |
| 1 g | |
| QZORVGYVDDOQNT-UHFFFAOYSA-N | |
| [3,5-bis(methoxymethoxy)phenyl]methanol | |
| 10443751 | |
| ≥98.0% (GC) | |
| 3,5-Bis(methoxymethyloxy)benzyl Alcohol |
Safety and Handling
TSCA : No