missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Bis(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
$249.78 - $249.78
Chemical Identifiers
| CAS | 725-89-3 |
|---|---|
| Molecular Formula | C9H4F6O2 |
| Molecular Weight (g/mol) | 258.12 |
| MDL Number | MFCD00000388 |
| InChI Key | HVFQJWGYVXKLTE-UHFFFAOYSA-N |
| Synonym | 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725 |
| PubChem CID | 12889 |
| IUPAC Name | 3,5-bis(trifluoromethyl)benzoic acid |
| SMILES | OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
Chemical Identifiers
| 725-89-3 | |
| 258.12 | |
| HVFQJWGYVXKLTE-UHFFFAOYSA-N | |
| 12889 | |
| OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F |
| C9H4F6O2 | |
| MFCD00000388 | |
| 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725 | |
| 3,5-bis(trifluoromethyl)benzoic acid |
Specifications
| 725-89-3 | |
| White-Yellow | |
| MFCD00000388 | |
| 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725 | |
| OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F | |
| 258.12 | |
| 258.12 | |
| Crystalline Powder |
| 142°C | |
| C9H4F6O2 | |
| 25 g | |
| HVFQJWGYVXKLTE-UHFFFAOYSA-N | |
| 3,5-bis(trifluoromethyl)benzoic acid | |
| 12889 | |
| ≥98.0% (GC,T) | |
| 3,5-Bis(trifluoromethyl)benzoic Acid |
Safety and Handling
RTECSNumber : DG4448020
TSCA : No