Learn More
3,5-Bis(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals
$133.63 - $517.11
Chemical Identifiers
CAS | 27126-93-8 |
---|---|
Molecular Formula | C9H3F6N |
Molecular Weight (g/mol) | 239.12 |
MDL Number | MFCD00000379 |
InChI Key | CZKHHAOIHXHOSR-UHFFFAOYSA-N |
Synonym | 3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene |
PubChem CID | 96746 |
IUPAC Name | 3,5-bis(trifluoromethyl)benzonitrile |
SMILES | FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1546006
|
Thermo Scientific Chemicals
A1546006 |
5 g |
Each for $133.63
|
|
|||||
AAA1546014
|
Thermo Scientific Chemicals
A1546014 |
25 g |
Each for $517.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27126-93-8 | |
239.12 | |
CZKHHAOIHXHOSR-UHFFFAOYSA-N | |
96746 | |
FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F |
C9H3F6N | |
MFCD00000379 | |
3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene | |
3,5-bis(trifluoromethyl)benzonitrile |
Specifications
27126-93-8 | |
1.421 | |
72°C (161°F) | |
1.4175 | |
5 g | |
3552650 | |
CZKHHAOIHXHOSR-UHFFFAOYSA-N | |
3,5-bis(trifluoromethyl)benzonitrile | |
96746 | |
98% |
16°C to 20°C | |
154°C to 156°C | |
C9H3F6N | |
MFCD00000379 | |
UN3276 | |
3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene | |
FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F | |
239.12 | |
239.12 | |
3,5-Bis(trifluoromethyl)benzonitrile |
Safety and Handling
GHS H Statement
H301-H227-H312-H332-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H227-H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 248-240-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only