missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl Bromide 96.0+%, TCI America™

$68.97 - $272.16

Chemical Identifiers

CAS 32247-96-4
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.03
MDL Number MFCD00009905
InChI Key ATLQGZVLWOURFU-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
PubChem CID 122573
IUPAC Name 1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18675G
View Documents
TCI America
B18675G
5 g
Each for $68.97
Only null left
 
B186725G
View Documents
TCI America
B186725G
25 g
Each for $272.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

32247-96-4
307.03
ATLQGZVLWOURFU-UHFFFAOYSA-N
122573
FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
C9H5BrF6
MFCD00009905
3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene

Specifications

32247-96-4
C9H5BrF6
5 g
3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
307.03
307.03
Liquid
140°C
MFCD00009905
2924
ATLQGZVLWOURFU-UHFFFAOYSA-N
1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
122573
≥96.0% (GC)
3,5-Bis(trifluoromethyl)benzyl Bromide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'