missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(5-bromo-2-thienyl)pyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 169050-05-9
Molecular Formula C9H6BrNS
Molecular Weight (g/mol) 240.12
MDL Number MFCD04971290
InChI Key GJBHIXTYLYHAQD-UHFFFAOYSA-N
Synonym 3-5-bromothiophen-2-yl pyridine, 3-5-bromo-2-thienyl pyridine, 3-5-bromothien-2-yl pyridine, 2-bromo-5-pyridin-3-ylthiophene, 5-bromo-2-3-pyridyl thiophene, 2-bromo-5-pyridin-3-yl thiophene
PubChem CID 818817
IUPAC Name 3-(5-bromothiophen-2-yl)pyridine
SMILES BrC1=CC=C(S1)C1=CN=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC66810CB
SDS View Documents
Thermo Scientific Chemicals
CC66810CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC66810DA
SDS View Documents
Thermo Scientific Chemicals
CC66810DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

169050-05-9
240.12
GJBHIXTYLYHAQD-UHFFFAOYSA-N
818817
BrC1=CC=C(S1)C1=CN=CC=C1
C9H6BrNS
MFCD04971290
3-5-bromothiophen-2-yl pyridine, 3-5-bromo-2-thienyl pyridine, 3-5-bromothien-2-yl pyridine, 2-bromo-5-pyridin-3-ylthiophene, 5-bromo-2-3-pyridyl thiophene, 2-bromo-5-pyridin-3-yl thiophene
3-(5-bromothiophen-2-yl)pyridine

Specifications

169050-05-9
100.0
Amber Glass Bottle
MFCD04971290
3-5-bromothiophen-2-yl pyridine, 3-5-bromo-2-thienyl pyridine, 3-5-bromothien-2-yl pyridine, 2-bromo-5-pyridin-3-ylthiophene, 5-bromo-2-3-pyridyl thiophene, 2-bromo-5-pyridin-3-yl thiophene
BrC1=CC=C(S1)C1=CN=CC=C1
240.12
240.12
3-(5-Bromo-2-thienyl)pyridine
96.5
97%
C9H6BrNS
250mg
GJBHIXTYLYHAQD-UHFFFAOYSA-N
3-(5-bromothiophen-2-yl)pyridine
818817
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'