Learn More
3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals
$77.62 - $222.79
Chemical Identifiers
CAS | 1421-49-4 |
---|---|
Molecular Formula | C30H42NiO6 |
Molecular Weight (g/mol) | 557.35 |
MDL Number | MFCD00008827 |
InChI Key | VIUVLVWUWLHYGT-UHFFFAOYSA-L |
Synonym | 3,5-di-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-9f0i7yag34, 3,5-di-t-butyl-4-hydroxybenzoic acid, 3,5-bis-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-di-tert-butyl-4-hydroxy, 3,5-di tert-butyl-4-hydroxybenzoic acid, 3,5-di-tert-butyl-4-hydroxy benzoic acid, 3,5-bis tert-butyl-4-hydroxybenzoic acid, pubchem15681 |
PubChem CID | 15007 |
IUPAC Name | 3,5-ditert-butyl-4-hydroxybenzoic acid |
SMILES | [Ni++].CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O.CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1688009
|
Thermo Scientific Chemicals
A1688009 |
10 g |
Each for $77.62
|
|
||||||
AAA1688018
|
Thermo Scientific Chemicals
A1688018 |
50 g |
Each for $222.79
|
|
||||||
Description
3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
Chemical Identifiers
1421-49-4 | |
557.35 | |
VIUVLVWUWLHYGT-UHFFFAOYSA-L | |
15007 | |
[Ni++].CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O.CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O |
C30H42NiO6 | |
MFCD00008827 | |
3,5-di-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-9f0i7yag34, 3,5-di-t-butyl-4-hydroxybenzoic acid, 3,5-bis-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-di-tert-butyl-4-hydroxy, 3,5-di tert-butyl-4-hydroxybenzoic acid, 3,5-di-tert-butyl-4-hydroxy benzoic acid, 3,5-bis tert-butyl-4-hydroxybenzoic acid, pubchem15681 | |
3,5-ditert-butyl-4-hydroxybenzoic acid |
Specifications
1421-49-4 | |
C30H42NiO6 | |
10 g | |
3,5-di-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-9f0i7yag34, 3,5-di-t-butyl-4-hydroxybenzoic acid, 3,5-bis-tert-butyl-4-hydroxybenzoic acid, benzoic acid, 3,5-di-tert-butyl-4-hydroxy, 3,5-di tert-butyl-4-hydroxybenzoic acid, 3,5-di-tert-butyl-4-hydroxy benzoic acid, 3,5-bis tert-butyl-4-hydroxybenzoic acid, pubchem15681 | |
VIUVLVWUWLHYGT-UHFFFAOYSA-L | |
3,5-ditert-butyl-4-hydroxybenzoic acid | |
15007 | |
98% |
206°C to 209°C | |
MFCD00008827 | |
2216948 | |
Insoluble in water. | |
[Ni++].CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O.CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C([O-])=O | |
557.35 | |
250.34 | |
3,5-Di-tert-butyl-4-hydroxybenzoic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 215-823-2
RTECSNumber : DG6320000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only