Learn More
3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture, Thermo Scientific Chemicals
$56.66 - $320.38
Chemical Identifiers
CAS | 535-87-5 |
---|---|
Molecular Formula | C7H8N2O2 |
Molecular Weight (g/mol) | 152.153 |
MDL Number | MFCD00007807 |
InChI Key | UENRXLSRMCSUSN-UHFFFAOYSA-N |
Synonym | benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid |
PubChem CID | 12062 |
IUPAC Name | 3,5-diaminobenzoic acid |
SMILES | C1=C(C=C(C=C1N)N)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1407518
|
Thermo Scientific Chemicals
A1407518 |
50 g |
Each for $56.66
|
|
|||||
AAA1407522
|
Thermo Scientific Chemicals
A1407522 |
100 g |
Each for $93.29
|
|
|||||
AAA1407536
|
Thermo Scientific Chemicals
A1407536 |
500 g |
Each for $320.38
|
|
|||||
Description
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
535-87-5 | |
152.153 | |
UENRXLSRMCSUSN-UHFFFAOYSA-N | |
12062 | |
C1=C(C=C(C=C1N)N)C(=O)O |
C7H8N2O2 | |
MFCD00007807 | |
benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid | |
3,5-diaminobenzoic acid |
Specifications
535-87-5 | |
210°C (410°F) | |
MFCD00007807 | |
2086484 | |
benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid | |
UENRXLSRMCSUSN-UHFFFAOYSA-N | |
3,5-diaminobenzoic acid | |
12062 | |
98% | |
may contain up to 3% moisture |
∼235°C (decomposition) | |
C7H8N2O2 | |
50 g | |
14,2978 | |
Very soluble in water. | |
C1=C(C=C(C=C1N)N)C(=O)O | |
152.153 | |
152.15 | |
3,5-Diaminobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-621-0
RTECSNumber : DG6186000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only