missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Diaminobenzoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 112111000

 View more versions of this product

Catalog No. AC112111000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3, 5-Diaminobenzoic acid
535-87-5
100.0
97.5% min. (HPLC)
C7H8N2O2
MFCD00007807
14, 453
benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid
UENRXLSRMCSUSN-UHFFFAOYSA-N
3,5-diaminobenzoic acid
12062
98%
98%
97.5
Authentic
Plastic bottle
(H2N)2C6H3CO2H
100g
15, 2980
Solubility in water: 7g/L (22°C). Other solubilities: soluble in alcohol and ether
C1=C(C=C(C=C1N)N)C(=O)O
152.15
152.15

Chemical Identifiers

535-87-5
152.15
UENRXLSRMCSUSN-UHFFFAOYSA-N
12062
C1=C(C=C(C=C1N)N)C(=O)O
C7H8N2O2
MFCD00007807
benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid
3,5-diaminobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-621-0

RTECSNumber : DG6186000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only