missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromo-4-methylphenol, 97%, Thermo Scientific Chemicals

$153.49 - $554.62

Chemical Identifiers

CAS 13979-81-2
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD00013968
InChI Key AXCQKKVGMZCWPC-UHFFFAOYSA-N
PubChem CID 55457
IUPAC Name 3,5-dibromo-4-methylphenol
SMILES CC1=C(C=C(C=C1Br)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0066003
View Documents
Thermo Scientific Chemicals
L0066003
1 g
Each for $153.49
 
AAL0066006
View Documents
Thermo Scientific Chemicals
L0066006
5 g
Each for $554.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13979-81-2
265.932
AXCQKKVGMZCWPC-UHFFFAOYSA-N
3,5-dibromo-4-methylphenol
C7H6Br2O
MFCD00013968
55457
CC1=C(C=C(C=C1Br)O)Br

Specifications

13979-81-2
C7H6Br2O
1 g
3,5-dibromo-p-cresol, phenol, 3,5-dibromo-4-methyl, 3,5-dibromo-4-methyl-phenol, phenol, dibromomethyl, acmc-20abwf, 3,5-dibromo-p-methylphenol, 3,5-dibromo-4-methyl phenol, phenol,3,5-dibromo-4-methyl, 3,5-bis bromanyl-4-methyl-phenol
CC1=C(C=C(C=C1Br)O)Br
265.932
265.94
3,5-Dibromo-4-methylphenol
106°C to 110°C
MFCD00013968
2249692
AXCQKKVGMZCWPC-UHFFFAOYSA-N
3,5-dibromo-4-methylphenol
55457
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-763-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only