missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%, Thermo Scientific Chemicals

$94.64 - $1642.49

Chemical Identifiers

CAS 54970-72-8
Molecular Formula C6H3Cl2NaO4S
Molecular Weight (g/mol) 265.036
MDL Number MFCD00009798
InChI Key NMWCVZCSJHJYFW-UHFFFAOYSA-M
Synonym sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
PubChem CID 3085077
IUPAC Name sodium;3,5-dichloro-2-hydroxybenzenesulfonate
SMILES C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1073209
View Documents
Thermo Scientific Chemicals
A1073209
10 g
Each for $94.64
 
AAA1073218
View Documents
Thermo Scientific Chemicals
A1073218
50 g
Each for $351.89
 
AAA1073230
View Documents
Thermo Scientific Chemicals
A1073230
250 g
Each for $1,642.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54970-72-8
265.036
NMWCVZCSJHJYFW-UHFFFAOYSA-M
3085077
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]
C6H3Cl2NaO4S
MFCD00009798
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
sodium;3,5-dichloro-2-hydroxybenzenesulfonate

Specifications

54970-72-8
C6H3Cl2NaO4S
10 g
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]
265.036
265.05
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt
>300°C
MFCD00009798
3640643
NMWCVZCSJHJYFW-UHFFFAOYSA-M
sodium;3,5-dichloro-2-hydroxybenzenesulfonate
3085077
99%
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 259-416-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only