missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichlorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1100306

 View more versions of this product

Catalog No. AAA1100306


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Dichlorobenzenesulfonyl chloride
30°C to 33°C
>110°C (230°F)
MFCD00051698
UN3261
Moisture sensitive
RJSQINMKOSOUGT-UHFFFAOYSA-N
3,5-dichlorobenzenesulfonyl chloride
522388
97%
705-21-5
158°C to 160°C (15 mmHg)
C6H3Cl3O2S
5 g
1528657
3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t
ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O
245.50
245.51

Chemical Identifiers

705-21-5
245.50
RJSQINMKOSOUGT-UHFFFAOYSA-N
522388
ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O
C6H3Cl3O2S
MFCD00051698
3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t
3,5-dichlorobenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only