missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichlorobenzonitrile, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1121303

 View more versions of this product

Catalog No. AAA1121303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Dichlorobenzonitrile
64°C to 68°C
MFCD00001800
UN3439
benzonitrile, 3,5-dichloro, 3,5-dichloro-benzonitrile, 3,5-dichlorobenzenecarbonitrile, pubchem4752, 3,5-dichlorobenzonitril, acmc-1b8n0, ksc354e0l, chembl51500, 3,5-dichlorobenzonitrile
ClC1=CC(=CC(Cl)=C1)C#N
172.01
172.01
6575-00-4
C7H3Cl2N
1 g
2207019
PUJSUOGJGIECFQ-UHFFFAOYSA-N
3,5-dichlorobenzonitrile
81052
98%

Chemical Identifiers

6575-00-4
172.01
PUJSUOGJGIECFQ-UHFFFAOYSA-N
81052
ClC1=CC(=CC(Cl)=C1)C#N
C7H3Cl2N
MFCD00001800
benzonitrile, 3,5-dichloro, 3,5-dichloro-benzonitrile, 3,5-dichlorobenzenecarbonitrile, pubchem4752, 3,5-dichlorobenzonitril, acmc-1b8n0, ksc354e0l, chembl51500, 3,5-dichlorobenzonitrile
3,5-dichlorobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 229-495-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only