Learn More
3,5-Dichlorophenol, 98%, Thermo Scientific Chemicals
$106.89 - $614.92
Chemical Identifiers
CAS | 591-35-5 |
---|---|
Molecular Formula | C6H4Cl2O |
Molecular Weight (g/mol) | 162.997 |
MDL Number | MFCD00002259 |
InChI Key | VPOMSPZBQMDLTM-UHFFFAOYSA-N |
Synonym | phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624 |
PubChem CID | 11571 |
IUPAC Name | 3,5-dichlorophenol |
SMILES | C1=C(C=C(C=C1Cl)Cl)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1532806
|
Thermo Scientific Chemicals
A1532806 |
5 g |
Each for $106.89
|
|
|||||
AAA1532814
|
Thermo Scientific Chemicals
A1532814 |
25 g |
Each for $369.67
|
|
|||||
AAA1532818
|
Thermo Scientific Chemicals
A1532818 |
50 g |
Each for $614.92
|
|
|||||
Description
3,5-Dichlorophenol is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dichlorophenol is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water. Very soluble in ethanol, ethyl ether and petroleum ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
591-35-5 | |
162.997 | |
VPOMSPZBQMDLTM-UHFFFAOYSA-N | |
11571 | |
C1=C(C=C(C=C1Cl)Cl)O |
C6H4Cl2O | |
MFCD00002259 | |
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624 | |
3,5-dichlorophenol |
Specifications
591-35-5 | |
233°C | |
C6H4Cl2O | |
5 g | |
1906458 | |
Slightly soluble in water. Very soluble in ethanol,ethyl ether and petroleum ether. | |
C1=C(C=C(C=C1Cl)Cl)O | |
162.997 | |
163 | |
3,5-Dichlorophenol |
63°C to 68°C | |
Phenol-like | |
MFCD00002259 | |
UN2020 | |
phenol, 3,5-dichloro, 3,5-dichloro-phenol, unii-fg32l88ko9, 3,5 dichlorophenol, ccris 5905, 3,5-dichloro phenol, pubchem3699, acmc-209mar, dsstox_cid_5006, dsstox_rid_77624 | |
VPOMSPZBQMDLTM-UHFFFAOYSA-N | |
3,5-dichlorophenol | |
11571 | |
98% |
Safety and Handling
GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber : 209-714-9
RTECSNumber : SK8820000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only