Learn More
3,5-Dichlorosalicylic acid, 99%, Thermo Scientific™
$41.12 - $41.12
Chemical Identifiers
CAS | 320-72-9 |
---|---|
Molecular Formula | C7H4Cl2O3 |
Molecular Weight (g/mol) | 207.006 |
MDL Number | MFCD00002442 |
InChI Key | CNJGWCQEGROXEE-UHFFFAOYSA-N |
Synonym | 3,5-dichlorosalicylic acid, benzoic acid, 3,5-dichloro-2-hydroxy, salicylic acid, 3,5-dichloro, usaf do-68, 2-hydroxy-3,5-dichlorobenzoic acid, unii-o6pxr32g3v, 3,5-dichlorosalicyclic acid, 3,5-dichlorosalicylicacid, 3,5-dichloro-2-hydroxy-benzoic acid, o6pxr32g3v |
PubChem CID | 9445 |
IUPAC Name | 3,5-dichloro-2-hydroxybenzoic acid |
SMILES | C1=C(C=C(C(=C1Cl)O)C(=O)O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2364106
|
Thermo Scientific Chemicals
B2364106 |
5 g |
Each for $41.12
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3,5-Dichlorosalicylic acid is a bio-active drug and is a potential inhibitor of human 20α-hydroxysteroid dehydrogenase. It also finds its application as medicine and dyes intermediates.
Solubility
Solubility in methanol (almost transparency).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
320-72-9 | |
207.006 | |
CNJGWCQEGROXEE-UHFFFAOYSA-N | |
9445 | |
C1=C(C=C(C(=C1Cl)O)C(=O)O)Cl |
C7H4Cl2O3 | |
MFCD00002442 | |
3,5-dichlorosalicylic acid, benzoic acid, 3,5-dichloro-2-hydroxy, salicylic acid, 3,5-dichloro, usaf do-68, 2-hydroxy-3,5-dichlorobenzoic acid, unii-o6pxr32g3v, 3,5-dichlorosalicyclic acid, 3,5-dichlorosalicylicacid, 3,5-dichloro-2-hydroxy-benzoic acid, o6pxr32g3v | |
3,5-dichloro-2-hydroxybenzoic acid |
Specifications
320-72-9 | |
C7H4Cl2O3 | |
5 g | |
CNJGWCQEGROXEE-UHFFFAOYSA-N | |
3,5-dichloro-2-hydroxybenzoic acid | |
9445 | |
99% |
222°C to 225°C | |
MFCD00002442 | |
3,5-dichlorosalicylic acid, benzoic acid, 3,5-dichloro-2-hydroxy, salicylic acid, 3,5-dichloro, usaf do-68, 2-hydroxy-3,5-dichlorobenzoic acid, unii-o6pxr32g3v, 3,5-dichlorosalicyclic acid, 3,5-dichlorosalicylicacid, 3,5-dichloro-2-hydroxy-benzoic acid, o6pxr32g3v | |
C1=C(C=C(C(=C1Cl)O)C(=O)O)Cl | |
207.006 | |
207.01 | |
3,5-Dichlorosalicylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : VO2450000
TSCA : TSCA